missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) isopropoxide, 99% (REO), 5% w/v in toluene/isopropanol, Thermo Scientific™

Catalog Number p-7051477
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
3504-40-3
C9H24O3Sm
330.648
MFCD00058820
BVFXJLVXFKLRNG-UHFFFAOYSA-N
samarium iii isopropoxide
85402317
propan-2-ol;samarium
CC(C)O.CC(C)O.CC(C)O.[Sm]
This item is not returnable. View return policy
3504-40-3
C9H24O3Sm
330.648
MFCD00058820
BVFXJLVXFKLRNG-UHFFFAOYSA-N
samarium iii isopropoxide
85402317
propan-2-ol;samarium
CC(C)O.CC(C)O.CC(C)O.[Sm]

Precursor for CVD and Sol-Gel

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Odor Alcohol-like
Linear Formula Sm[OCH(CH3)2]3
Quantity 25 mL
UN Number UN3274
Sensitivity Moisture Sensitive
Formula Weight 327.5
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Samarium(III) isopropoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'