missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Samarium foil, 0.75mm (0.03 in.) thick, 99.9% (REO)

Brand:  Alfa Aesar™ 043218.FI

Code : NEW

Additional Details : CAS Number : 7440-19-9 Weight : 0.01000kg

Quantity 50 x 50mm

 View more versions of this product

Product Code. 11306209

  • £538.80 / Each


Estimated Shipment date
from Supplier 05-03-2018
Add to basket

Specifications

Chemical Identifiers

Samarium foil
7440-19-9
0.75mm (0.03 in.) thick
MFCD00011233

Specifications

Samarium foil
50 x 50mm
7440-19-9
Mylar under Argon
99.9%
MFCD00011233
≈3.7g/25x25mm
0.75mm (0.03 in.) thick
Odorless
Foil
(REO)
Videos
SDS