missing translation for 'onlineSavingsMsg'
Learn More

Samarium foil, 0.25mm (0.01in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number p-7049000
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
This item is not returnable. View return policy
7440-19-9
Sm
150.36
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]
This item is not returnable. View return policy
7440-19-9
Sm
150.36
MFCD00011233 MFCD00151299
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
23951
CHEBI:33374
samarium
[Sm]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Quantity 25 x 25 mm
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈1.2g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Samarium foil, 0.25mm (0.01 in.) thick

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'