missing translation for 'onlineSavingsMsg'
Learn More

Samarium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

Catalog Number 15436125
Click to view available options
Quantity:
25 x 25 mm
Packaging:
Mylar under Argon
This item is not returnable. View return policy
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY
Color Silver
CAS 7440-19-9
Molecular Formula Sm
MDL Number MFCD00011233 MFCD00151299
UN Number UN3178
Quantity 25 x 25 mm
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES [Sm]
IUPAC Name samarium
Molecular Weight (g/mol) 150.36
PubChem CID 23951
ChEBI CHEBI:33374
Percent Purity 99.9%
Odor Odorless
Packaging Mylar under Argon
Weight ≈0.12g/25x25mm
Form Foil
Assay (REO)
Chemical Name or Material Samarium foil, 0.025mm (0.001 in.) thick
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'