missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Salubrinal
Click to view available options
Quantity:
5 mg
10 mg
Description
Salubrinal is a selective Inhibitor of eIF2α (eukaryotic translation initiation factor 2 α-subunit) dephosphorylation, inhibiting global protein translation. Salubrinal is a specific inhibitor of ER Stress induced apoptosis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 5 mg |
Description | Pale Brown |
Content And Storage | 2°C to 8°C |
Formula Weight | 479.81g/mol |
Sensitivity | Light Sensitive |
Product Type | Apoptosis Reagent |
Form | Powder |
InChI Key | LCOIAYJMPKXARU-UHFFFAOYNA-N |
SMILES | ClC(Cl)(Cl)C(NC(=S)NC1=C2N=CC=CC2=CC=C1)NC(=O)C=CC1=CC=CC=C1 |
IUPAC Name | 3-phenyl-N-(2,2,2-trichloro-1-{[(quinolin-8-yl)carbamothioyl]amino}ethyl)prop-2-enamide |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'