missing translation for 'onlineSavingsMsg'
Learn More

Saline-Sodium Phosphate-EDTA (SSPE), 20X Solution, Fisher BioReagents

Brand:  Fisher Bioreagents BP1328-20

Code : 32

Additional Details : Weight : 20.05000kg

Quantity 20L
Packaging Poly CUBE

 View more versions of this product

Product Code. 10499303

  • £161.80 / 20 L


Estimated Shipment
16-03-2018
Add to basket

Specifications

Chemical Identifiers

Saline-Sodium Phosphate-EDTA
20X Solution
7732-18-5
SSPE Buffer

Specifications

Saline-Sodium Phosphate-EDTA
7732-18-5
2.9 to 3.1M
SSPE Buffer
Poly CUBE
Filtered through a 2-5 micron filter.
20L
20X Solution
Liquid
DNase free
Sodium Chloride (17.5%), Sodium Biphosphate (2.8%), and Ethylenediamine Tetraacetic Acid (0.6%).
Protease free
Clear
Videos
SDS Documentation