missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Salicylanilide, 98%

Brand:  Alfa Aesar™ A10335.22

Code : NEW

Additional Details : CAS Number : 87-17-2 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11454113

  • £13.00 / 100g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Description

Description

Applications
It is employed as a drug used to treat rheumatoid arthritis and has principally analgesic, antipyretic and anti-inflammatory actions. Non-narcotic analgesics do not bind to opioid receptors. It is used mainly in medicated soaps.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Chemical Identifiers

Salicylanilide
C13H11NO2
MFCD00002212
2-Hydroxybenzanilide
87-17-2
213.24
14,8330

Specifications

Salicylanilide
98%
213.24
14,8330
136° to 138°C
87-17-2
C13H11NO2
MFCD00002212
2-Hydroxybenzanilide
100g
Videos
SDS