Learn More
SALF Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552190
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DSPLAISGGE SSLLPTRPTC LSHALLPSDH SCTHPTPKVG LPDEVNPQQA ESLGFQSDDL PQFQYFREDC AFSSPFWKDE GSDSHFTLDP PGSKKMFSSR NKEMPIDQKS LNKCSLNYIC EKLEHLQSAE NQDSLR Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 54%.
SALF Polyclonal antibody specifically detects SALF in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SALF | |
Polyclonal | |
Unconjugated | |
STON1 | |
4921524J06Rik; C86231; Salf; Sblf; STN1; STNB1; STON1; STON1-GTF2A1L; STON1-GTF2A1L protein; STON1-GTF2A1L readthrough; stoned B homolog 1; stoned B/TFIIA-alpha/beta-like factor; stoned B-like factor; stoned-b1; stonin 1; Stonin1; stonin-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y6Q2 | |
STON1 | |
Recombinant protein corresponding to Human SALF | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only