missing translation for 'onlineSavingsMsg'
Learn More

Salermide, Tocris Bioscience™

SIRT1 and SIRT2 inhibitor

Brand:  Tocris (Bio-Techne) 412750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13434204

  • £311.64 / 50mg

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SIRT1 and SIRT2 inhibitor. Exhibits a stronger inhibitory effect on SIRT2 than on SIRT1 in vitro. Induces the reactivation of proapoptotic genes repressed by SIRT1 and causes massive apoptosis in cancer cells within 24 hours.
Specifications

Chemical Identifiers

1105698-15-4
394.474
N-[3-[[(2-Hydroxy-1-naphthalenyl)methylene]amino]phenyl]-α-methylbenzeneacetamide
(2S)-N-[3-[[(E)-(2-oxonaphthalen-1-ylidene)methyl]amino]phenyl]-2-phenylpropanamide
C26H22N2O2
UADRPWLRVLBVBC-KHCKMINHSA-N
98114484
CC(C1=CC=CC=C1)C(=O)NC2=CC=CC(=C2)NC=C3C(=O)C=CC4=CC=CC=C43

Specifications

50mg
1105698-15-4
N-[3-[[(2-Hydroxy-1-naphthalenyl)methylene]amino]phenyl]-α-methylbenzeneacetamide
CC(C1=CC=CC=C1)C(=O)NC2=CC=CC(=C2)NC=C3C(=O)C=CC4=CC=CC=C43
394.474
394.47
Salermide
C26H22N2O2
UADRPWLRVLBVBC-KHCKMINHSA-N
(2S)-N-[3-[[(E)-(2-oxonaphthalen-1-ylidene)methyl]amino]phenyl]-2-phenylpropanamide
98114484
>98%
Videos
Documents