Learn More
Tocris Bioscience™ Sal 003
Cell-permeable inhibitor of eIF2α dephosphorylation
Brand:  Tocris Bioscience™ 365710
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13463364
  • £124.20 / 10mg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Cell-permeable inhibitor of cellular phosphatase complexes that dephosphorylate eukaryotic translation initiation factor 2 subunit α (eIF2α). Analog of salubrinal (Cat. No. 2347) with improved aqueous solubility. Shown to prevent the induction of hippocampal long-term potentiation (LTP) and memory formation (LTM) in mice.
Specifications
Specifications
Sal 003
>99
10mg
Sal 003
463.21