Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
Brand: Saint-Gobain MFLX96430-04 (EA)
Code : A0
Additional Details : Weight : 0.80000kg
Description
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
- Does not contaminate fluids
- Superior flex life in peristaltic pumps
- Chemically resistant to a wide range of fluids
- Crystal clear for easy visual flow monitoring
- Meets FDA criteria for food contact
- Will not degrade over time
- Will not release hazardous byproducts when properly incinerated
- Does not yield any taste
- Helps ensure accurate results from analytical instrument tests
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
†At 73°F (23°C).
Specifications
Compliant with FDA 21 CFR, 177.2600 | |
4.8 mm | |
7.9 mm | |
1/16 in. |
3/16 in. | |
5/16 in. | |
50 ft. | |
1.6 mm |