missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain™ Tygon™ E-1000 Ultra-Soft Tubing

Non-DEHP Laboratory Tubing

Brand:  Saint-Gobain™ ADK00027

Code : 02

Additional Details : Weight : 1.67000kg

 View more versions of this product

Product Code. 13193603

  • £162.09 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

9.5mm
12.7mm
15m
Tygon
50 ft.
1.6mm
Videos