missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Tygon™ E-1000 Tubing

Flexible, food and beverage dispensing Tygon™ E-1000 Tubing.

Brand:  Saint Gobain Life Sciences

Code : 02

Additional Details : Weight : 2.11000kg

 View more versions of this product

Product Code. 13113613

  • £540.00 / Each
Estimated Shipment: 13-09-2024
to see stock.
Add to basket

Description

Description

Soft and flexible, Tygon™ E-1000 tubing delivers superior performance in a formulation that contains non-DEHP [Bis (2-ethylhexyl) phthalate] plasticizers. Tygon™ E-1000 non-DEHP tubing has been tested rigorously to meet stringent standards for low temperature and corrosive chemical resistance.

  • Ultra-smooth inner bore reduces potential for particle entrapment
  • Minimal extractable help maintain fluid integrity
  • Excellent fluid flow characteristics
  • Complete inventory of standard sizes available, including metric sizes

Typical Applications

  • Food and beverage dispensing
Specifications

Specifications

Tubing
12.7 mm
0.75 in.
50 ft.
1/8 in.
-55°C
8 psi
52°C
0.5 in.
Tygon E-1000
19 mm
15 m
3.2 mm
40 (Shore A)
178 mm Hg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers