missing translation for 'onlineSavingsMsg'
Learn More

Saint-Gobain™ Chemware™ PTFE Fluoropolymer Resin Policemen

For delicate transfer or scraping of precipitates and residues

Brand:  Saint-Gobain™ D1075014

Code : 02

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 11970495

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Description

Description

  • Will not scratch or damage delicate surfaces
  • Choose from Hayman Style, Double Flat End Style, Double-ended Lab Spoon Style and Tapered Pallet with reinforced Hardwood Handle
  • Extra heavy-duty PTFE coatings
  • Resistant to low and high temperatures
  • Use for handling chemically active materials
  • Corrosion resistant
  • Non-wettable
  • Chemically inert
Specifications

Specifications

Spatula, Policemen
0.023cm
PTFE Fluoropolymer Resin
Delicate transfer or scraping of precipitates and residues
Chisel tip and diagonal cut end
20cm
Videos