missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Chemware™ Bendable PTFE Stirring Rod

Chemware™ Bendable PTFE Stirring Rod contains a fully enclosed steel insert for stiffness, yet can be bent to various shapes for use as powered stirrers.

Brand:  Saint Gobain Life Sciences D1069091

Code : 03

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12348219

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Heat resistant to 288°C (550°F)
  • Completely inert to highly corrosive reagents
  • Anti-stick properties facilitate cleaning
  • Unbreakable
Specifications

Specifications

Fully enclosed steel insert
16 in.
Stirring Rod
PTFE
8 mm
0.3 in.
1 Ea.
40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers