Learn More
SCAT SafetyWasteCap S70/71 Thread
Safely connect your system waste tubes directly to your disposal system container via the ports in this cap.
Brand:  SCAT 108407
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 10783302
  • £384.23 / Each
Call for latest availability
+44 (0)1509 555 500
Description
Description
  • Variety of waste cap styles available with different connection arrays available
  • Designed to provide fittings and accessories for safe waste collection
  • For containers with S70/71 threads
Specifications
Specifications
Includes 2 x 2.3/3.2 mm OD capillary connectors, 1 x 6.4-9.0 mm OD tube connector, 1 x connector for exhaust filter, 1 x electronic level control
Waste disposal system
Waste disposal container with S70/71 thread opening
Waste Disposal System Cap