Learn More
SCAT SafetyWasteCap, S40/GL40 Thread
Safely connect your system waste tubes directly to your disposal system container via the ports in this cap.
Brand:  SCAT 107108
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 12603335
Quantity Price
1 £83.49 / Each
10 £78.65 / Each
Estimated Shipment
25-09-2020
Add to basket
Description
Description
  • Variety of waste cap styles available with different connection arrays available
  • Designed to provide fittings and accessories for safe waste collection
  • For containers with S40/GL40 thread openings
Specifications
Specifications
Includes 3 x capillary connectors 2.3/3.2 mm OD, and 1 x connector for exhaust filter
Waste containers with S40/GL40 thread openings.