Learn More
Sartorius SafetySpace™ Filter Pipet Tips
Provides more space between the sample and the filter than conventional filter tips which virtually eliminates the risk of an expensive or a contagious sample permeating the filter. Sartorius SafeySpace™ Filter Pipet Tips allow pipetting any type of liquid or using any pipetting technique without the risk of the sample absorbing into the filter.
£195.00 - £326.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Autoclavable | Not Autoclavable |
Certifications/Compliance | CE, ISO, IVD |
Clean Claims | DNase, RNase, and Endotoxin Free |
Product Code | Brand | Color Code | Length (Metric) | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color Code | Length (Metric) | Volume (Metric) | Price | Quantity & Availability | |||
10587600
|
Sartorius
790011F |
Gray | 3.2cm | 0.1 to 10 μL |
£286.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
13197873
|
Sartorius
790101F |
Yellow | 5.1cm | 2 to 120 μL |
£288.00
Pack of 960 |
Estimated Shipment: 01-05-2024
Log in to see stock. |
|||
10014554
|
Sartorius
790201F |
Yellow | 5.25cm | 5 to 200 μL |
£284.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
13107634
|
Sartorius
790301F |
Orange | 5.25cm | 5 to 300 μL |
£288.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
11894782
|
Sartorius
791211F |
Purple | 9cm | 50 to 1200 μL |
£326.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
10708262
|
Sartorius
783201 |
Gray | 4.6cm | 0.1 to 10 μL |
£195.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
11552934
|
Sartorius
791001F |
Blue | 7.8cm | 5 to 1000 μL |
£288.00
Pack of 960 |
In Stock
Log in to see stock. |
|||
Description
SafetySpace™ Filter Tips leave more space between the sample and the filter than conventional filter tips. This allows pipetting any type of liquid or using any pipetting technique without the risk of the precious sample absorbing into the filter. In effect, this secures 100% sample recovery and thus greater accuracy.- Polypropylene
- With regular air gap and filter barrier
- Fully autoclavable
- Single tray packaging
- Purity level: RNase, DNase, Pre-endotoxin-free sterilized
The safety space feature is particularly useful in:
- pipetting foaming liquids such as buffers and proteins
- pipetting solvents
- multiple dispensing functions of electronic pipettes
- reverse pipetting
- any other application requiring extra space in the tip
The tips are made of virgin polypropylene and the filter of polyethylene without "self-sealing" additives to avoid any interference with the sample and the results. The filter barrier helps protect against cross-contamination of the sample and/or the pipette and reduces the need for pipette maintenance. The filter tips are available in multiple volumes from 10 μl to 1200 μl and are packed in colour coded single tray racks. They are DNase, RNase and endotoxin free certified and pre-sterilized.
The filter barrier and the certified purity makes the filter tips ideal for:
- molecular biology
- microbiology
- cell culture applications
- radioactive work
Specifications
Filtered | |
Not Autoclavable | |
DNase, RNase, and Endotoxin Free | |
Rack |
Sterile | |
CE, ISO, IVD | |
Pre-sterilized | |
Polypropylene |