Allsigns International™ Safety Signs Danger Fragile Roof


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA47/PVC.

Safety sign WA47/PVC hazard warning dangerfragile roof 200mm x 300mm 1mm rigid plastic

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385618

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
02-03-2017
Add to basket


Description and Specification

Specification

Material 1mm rigid plastic
Message Danger fragile roof
Quantity 1Each
Type Safety Signs
Adhesive Type PVC
Dimensions (L x W) 200 x 300mm