Allsigns International™ Safety Signs Danger Fragile Roof


Overview
Brand: Allsigns International™

Manufacturer Part Number: WA47/SA.

Safety sign WA47/SA hazard warning danger fragileroof 200mm x 300mm self adhesive vinyl Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12375618

Quantity Price
1 £ 11.97 / Each
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

Material Self adhesive vinyl
Message Danger fragile roof
Quantity 1Each
Type Safety Signs
Adhesive Type Self adhesive
Dimensions (L x W) 200 x 300mm