Allsigns International™ Safety Signs Danger Drop Below


Overview
Brand: Allsigns International™

Manufacturer Part Number: EC25/PVC.

Safety sign EC25/PVC hazard warning danger dropbelow 200mm x 300mm 1mm rigid plastic Allsigns,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12385698

Quantity Price
1 £ 21.24 / Each
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

Material 1mm rigid plastic
Message Danger drop below
Quantity 1Each
Type Safety Signs
Adhesive Type PVC
Dimensions (L x W) 200 x 300mm
Documentation

Technical documents