ALLSIGNS International™ Safety Sign


Overview
Brand: ALLSIGNS International™

Manufacturer Part Number: SA27/SA.

Safety sign SA27/SA fire exit with running man300mm x 150mm self adhesive vinyl Allsigns, 1/Cs,

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12349817

Quantity Price
1 £ 9.37 / Each
Estimated Shipment
04-04-2017
Add to basket


Description and Specification

Specification

Size (Metric) 300 x 250mm
Quantity 1Each
Type Safety sign
Material Vinyl
Message Fire Exit with Running Man
Documentation

Technical documents