ALLSIGNS International™ Safety Sign


Overview
Brand: ALLSIGNS International™

Manufacturer Part Number: PH13

Safety sign PH13. photoluminescent fire exit signc/w running man Gemglow self adhesive backed 1mm

UNSPSC: 55121704

Code: 04

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 12319857

Quantity Price
1 £ 36.93 / Each
Estimated Shipment
03-04-2017
Add to basket


Description and Specification

Specification

Size (Metric) 400 x 125mm
Quantity 1Each
Type Safety sign
Material Rigid Plastic
Message Fire Exit
Documentation

Technical documents