Learn More
Kimberly Clark™ safety Nemesis™ eyewear.
Brand:  Kimberly Clark™ 25697
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12671406
  • £53.30 / Pack of 12
Estimated Shipment
12-10-2017
Add to basket
Specifications
Specifications
Safety Eyewear
Reducing glare and keeping light levels constant when moving between natural and artificial light, staying in place during intense, active use
0.264kg
1/ea.
EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity.