Kimberly Clark™ safety eyewear.
Overview
Brand: Kimberly Clark™
Manufacturer Part Number: 25644
X12 EYEWEAR V10 ELEMENT IN/OUT
UNSPSC: 41120000
Code: NEW
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 12681396
Quantity Price
1 £ 32.0 / Pack of 12
Estimated Shipment
09-03-2017
Add to basket
Description and Specification
Specification
Quantity 12/Pack
Weight (Metric) 0.336kg
Product Type Safety Eyewear
For Use With (Application) Providing workers and visitors with basic eye protection
Certifications/Compliance EN166:2001 1F for safety, EN166:2001 level F for impact resistance and EN166:2001 class 1 for optical clarity