Learn More
Saclofen, Tocris Bioscience™
Selective GABAB antagonist
Brand:  Tocris (Bio-Techne) 024650
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487503
  • £674.00 / 50mg
Estimated Shipment
07-03-2018
Add to basket
Description
Description
Selective antagonist at GABAB receptors.
Specifications
Chemical Identifiers
Saclofen
C9H12ClNO3S
249.72
(RS)-3-Amino-2-(4-chlorophenyl)propylsulfonic acid
Specifications
Saclofen
>99%
249.72
50mg
125464-42-8
C9H12ClNO3S
(RS)-3-Amino-2-(4-chlorophenyl)propylsulfonic acid
Videos