missing translation for 'onlineSavingsMsg'
Learn More

Saccharin, 98+%, Thermo Scientific Chemicals

Catalog Number p-4510099
15.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
81-07-2
C7H5NO3S
183.18
CVHZOJJKTDOEJC-UHFFFAOYSA-N
saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose
5143
CHEBI:32111
1,1-dioxo-1,2-benzothiazol-3-one
C1=CC=C2C(=C1)C(=O)NS2(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
81-07-2
C7H5NO3S
183.18
CVHZOJJKTDOEJC-UHFFFAOYSA-N
saccharin, saccharine, o-sulfobenzimide, o-benzoic sulfimide, saccharimide, benzosulfimide, o-benzosulfimide, benzoic sulfimide, benzosulphimide, saccharinose
5143
CHEBI:32111
1,1-dioxo-1,2-benzothiazol-3-one
C1=CC=C2C(=C1)C(=O)NS2(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 226.0°C to 230.0°C
Color White
Loss on Drying 1% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Plastic bottle
Quantity 100 g
Beilstein 27,168
Merck Index 15,8445
Solubility Information Solubility in water: 3.3g/L (20°C). Other solubilities: 1g/31mL alcohol - 1g/12mL acetone, 1g/50mL glycerol, soluble in amyl acetate and ethyl acetate, slightly soluble in chloroform and ether, soluble in dil. solution of ammonia
Formula Weight 183.18
Percent Purity 98+%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Saccharin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'