missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura™ Electro 926 Micropipette

Lightweight and ergonomically friendly, this electronic single-channel micropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 926 Micropipette charges quickly and has a long working autonomy.

Brand:  Socorex™ 926.0002

Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11809213

  • £550.00 / Each
Estimated Shipment: 21-06-2024
to see stock.
Add to basket

Description

Description

  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Reduced shaft length and conical end improve drivability and access to microtubes and microplates.
  • Includes pipetting cycle counter.
  • Optional volumetric modules.
Specifications

Specifications

<±2.5% - <±0.9%
EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
Acura™ Electro™ 926
Acura™ Electro™
Electronic Micropipette
Right/Left LCD
0.01μL
1 Each
24 Months
4.8V NiMH
Pipette
1
Self-Teaching
0.1 to 2.0 μL
QC certificate, Qualitips™ pipette tip samples and operating instructions.
<2.5% - <0.8%
<1.5Hours
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers