missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ S100A8 Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human S100A8 (Leu 2-Glu 93) fused to polyhistidine tag at N-terminus

Brand:  Thermo Scientific™ 11138H08B50

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15288275

  • £216.53 / 50µg


Estimated Shipment
27-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

  • Measured by its ability to bind recombinant human S100A9 in functional ELISA
  • Measured by its ability to inhibit t proliferation of MCF7 human breast adenocarcinoma cells
  • ED50 for effect is 3-15μg/mL
  • Lyophilized in 140mM
Specifications

Specifications

Lyophilized
≥90% by SDS-PAGE
Human
<1 EU/μg
S100A8
Other proteins
S100A8 Recombinant Human Protein
12.2kd
50μg
Recombinant
S100 Calcium Binding Protein A8
Assay Standard
Insect
His Tag, MBP
Videos