missing translation for 'onlineSavingsMsg'
Learn More

(S)-WAY 100135 dihydrochloride, Tocris Bioscience™

Potent, selective 5-HT1A antagonist

Brand:  Tocris (Bio-Techne) 125350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458983

  • £557.72 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Potent, selective 5-HT1A receptor antagonist (IC50 = 15 nM). Selective over 5-HT1B, 1C, 2,α1, α2 and D2 receptors (IC50 > 1000 nM). Centrally active on systemic administration.
Specifications

Chemical Identifiers

149007-54-5
468.463
s-n-tert-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-way 100135 dihydrochloride, s-n-t-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, 2s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride
(2S)-N-tert-butyl-3-[4-(2-methoxyphenyl)piperazin-1-yl]-2-phenylpropanamide;dihydrochloride
C24H35Cl2N3O2
VJGZNBYDSDEOED-FAVHNTAZSA-N
14801907
CC(C)(C)NC(=O)C(CN1CCN(CC1)C2=CC=CC=C2OC)C3=CC=CC=C3.Cl.Cl

Specifications

(S)-WAY 100135 dihydrochloride
149007-54-5
s-n-tert-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-way 100135 dihydrochloride, s-n-t-butyl-3-4-2-methoxyphenyl-piperazin-1-yl-2-phenylpropanamide dihydrochloride, 2s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride, s-n-tert-butyl-3-4-2-methoxyphenyl piperazin-1-yl-2-phenylpropanamide dihydrochloride
CC(C)(C)NC(=O)C(CN1CCN(CC1)C2=CC=CC=C2OC)C3=CC=CC=C3.Cl.Cl
468.463
468.47
50mg
C24H35Cl2N3O2
VJGZNBYDSDEOED-FAVHNTAZSA-N
(2S)-N-tert-butyl-3-[4-(2-methoxyphenyl)piperazin-1-yl]-2-phenylpropanamide;dihydrochloride
14801907
>99%
Videos