missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Tetrahydro-3-furoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7057062
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
168395-26-4
C5H8O3
116.12
MFCD07369984
BOTREHHXSQGWTR-UHFFFAOYNA-N
s-tetrahydrofuran-3-carboxylic acid, 3s-oxolane-3-carboxylic acid, s-tetrahydro-3-furancarboxylic acid, s-tetrahydro-3-furoic acid, s-tetrahydro-furan-3-carboxylic acid, 3s-tetrahydrofuran-3-carboxylic acid, l-tetrahydrofuran-3-carboxylic acid, 3-furancarboxylic acid, tetrahydro-, 3s, 3s-3-oxolanecarboxylic acid, s, ?-tetrahydro-3-furancarboxylic acid
40784874
oxolane-3-carboxylic acid
OC(=O)C1CCOC1
This item is not returnable. View return policy
168395-26-4
C5H8O3
116.12
MFCD07369984
BOTREHHXSQGWTR-UHFFFAOYNA-N
s-tetrahydrofuran-3-carboxylic acid, 3s-oxolane-3-carboxylic acid, s-tetrahydro-3-furancarboxylic acid, s-tetrahydro-3-furoic acid, s-tetrahydro-furan-3-carboxylic acid, 3s-tetrahydrofuran-3-carboxylic acid, l-tetrahydrofuran-3-carboxylic acid, 3-furancarboxylic acid, tetrahydro-, 3s, 3s-3-oxolanecarboxylic acid, s, ?-tetrahydro-3-furancarboxylic acid
40784874
oxolane-3-carboxylic acid
OC(=O)C1CCOC1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow to Brown
Quantity 1 g
Beilstein 1281203
Formula Weight 116.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(-)-Tetrahydro-3-furoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'