missing translation for 'onlineSavingsMsg'
Learn More

(S)-tert-Leucinol, 95%, Thermo Scientific Chemicals

Catalog Number p-3776676
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
112245-13-3
C6H16NO
118.20
MFCD00192250
JBULSURVMXPBNA-RXMQYKEDSA-O
l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
2734079
CC(C)(C)[C@H]([NH3+])CO
This item is not returnable. View return policy
112245-13-3
C6H16NO
118.20
MFCD00192250
JBULSURVMXPBNA-RXMQYKEDSA-O
l-tert-leucinol, s-tert-leucinol, 2s-2-amino-3,3-dimethylbutan-1-ol, 2s-2-amino-3,3-dimethyl-butan-1-ol, s-2-amino-3,3-dimethylbutanol, s-2-amino-3,3-dimethyl-1-hydroxybutane, r---tert-leucinol, 1-butanol, 2-amino-3,3-dimethyl-, 2s
2734079
CC(C)(C)[C@H]([NH3+])CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Infrared Spectrum Authentic
Color White or Colorless
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Specific Rotation +34° to +40° (25°C, 589nm) (c=1.5, EtOH)
Quantity 250 mg
Physical Form Crystalline Low Melting Mass or Liquid
Assay Percent Range 95%
Linear Formula (CH3)3CCH(NH2)CH2OH
IUPAC Name (2S)-2-amino-3,3-dimethylbutan-1-ol
Formula Weight 117.19
Percent Purity ≥94%
Chemical Name or Material (S)-tert-Leucinol
Show More Show Less
compliance-icons
Product Identifier
  • (S)-tert-Leucinol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'