missing translation for 'onlineSavingsMsg'
Learn More

(S)-tert-Butylsulfinamide, 97+%, (>98.0% ee), Thermo Scientific Chemicals

Catalog Number p-3598221
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
343338-28-3
C4H11NOS
121.2
MFCD05861480
CESUXLKAADQNTB-ZETCQYMHSA-N
s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
11355477
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N
This item is not returnable. View return policy
343338-28-3
C4H11NOS
121.2
MFCD05861480
CESUXLKAADQNTB-ZETCQYMHSA-N
s---2-methyl-2-propanesulfinamide, s-2-methylpropane-2-sulfinamide, s-tert-butanesulfinamide, s---tert-butanesulfinamide, s---t-butylsulfinamide, s---tert-butylsulfinamide, s-2-methyl-2-propanesulfinamide, s-tert-butylsulfinamide, s---t-butylmethylsulfinamide, s-t-butyl sulfinamide
11355477
2-methylpropane-2-sulfinamide
CC(C)(C)S(=O)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Melting Point 101°C
Color White
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSONH2
Quantity 1 g
Merck Index 15, 3607
Formula Weight 121.2
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material (S)-tert-Butylsulfinamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'