missing translation for 'onlineSavingsMsg'
Learn More

S-tert-Butyl-L-cysteine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number 15485488
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2481-09-6
MFCD00037426
This item is not returnable. View return policy
2481-09-6
MFCD00037426

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Molecular Formula C10H16Si2
Synonym 1,4-bis dimethylsilyl benzene, benzene, 1,4-bis dimethylsilyl, p-phenylenebis dimethylsilane, 1,4-phenylenebis dimethylsilane, p-bis dimethylsilyl benzene, silane, p-phenylenebis dimethyl, silane, 1,4-phenylenebis dimethyl, 1,4 bis dimethylsilyl benzene
InChI Key UHXCHUWSQRLZJS-UHFFFAOYSA-N
SMILES C[Si](=C1C=CC(=[Si](C)C)C=C1)C
IUPAC Name (4-dimethylsilylidenecyclohexa-2,5-dien-1-ylidene)-dimethylsilane
Molecular Weight (g/mol) 192.41
PubChem CID 6328729
Formula Weight 213.73
Percent Purity 98%
Quantity 5 g
Physical Form Powder
Chemical Name or Material S-tert-Butyl-L-cysteine hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'