Learn More
Tocris Bioscience™ S-Sulfo-L-cysteine sodium salt
Brand:  Tocris Bioscience™ 016210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13487443
  • £173.00 / 10mg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
EAA receptor agonist. Agonist at mGlu1α and mGlu5a subtypes expressed in clonal RGT cell lines.
Specifications
Specifications
7381-67-1
Observed MW: 223.19
Soluble to 100 mM in water
C3H6NO5S2Na