missing translation for 'onlineSavingsMsg'
Learn More

(S)-Styrene oxide, 94%, Thermo Scientific Chemicals

Catalog Number p-4812530
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
20780-54-5
C8H8O
120.15
MFCD00064310,MFCD00066210
AWMVMTVKBNGEAK-MRVPVSSYSA-N
s-styrene oxide, s-2-phenyloxirane, 2s-2-phenyloxirane, s-phenyloxirane, s-epoxyethyl benzene, s-+-styrene oxide, styrene oxide, s, unii-av5p894c84, ccris 4094
114946
CHEBI:51014
(2S)-2-phenyloxirane
C1O[C@H]1C1=CC=CC=C1
This item is not returnable. View return policy
20780-54-5
C8H8O
120.15
MFCD00064310,MFCD00066210
AWMVMTVKBNGEAK-MRVPVSSYSA-N
s-styrene oxide, s-2-phenyloxirane, 2s-2-phenyloxirane, s-phenyloxirane, s-epoxyethyl benzene, s-+-styrene oxide, styrene oxide, s, unii-av5p894c84, ccris 4094
114946
CHEBI:51014
(2S)-2-phenyloxirane
C1O[C@H]1C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -37.0°C
Density 1.0510g/mL
Boiling Point 192.0°C to 194.0°C
Flash Point 79°C
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.051
Solubility Information Solubility in water: practically insoluble
Formula Weight 120.15
Percent Purity 94%
Chemical Name or Material (S)-Styrene oxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-Styrene oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'