missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-sec-Butanol, 99%, Thermo Scientific Chemicals

Catalog Number p-77949
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4221-99-2
C4H10O
74.12
MFCD00064281
BTANRVKWQNVYAZ-BYPYZUCNSA-N
s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
444683
CHEBI:45475
(2S)-butan-2-ol
CCC(C)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
4221-99-2
C4H10O
74.12
MFCD00064281
BTANRVKWQNVYAZ-BYPYZUCNSA-N
s-+-2-butanol, s-2-butanol, 2s-butan-2-ol, s-butan-2-ol, s-+-sec-butanol, 2s-2-butanol, unii-69kxu5ndto, s---2-butanol, s-+-sec-butyl alcohol
444683
CHEBI:45475
(2S)-butan-2-ol
CCC(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -114°C
CAS Min % 98.5
Color Colorless
CAS Max % 100
Density 0.8000g/mL
Boiling Point 99°C to 100°C
Quantity 1 g
Flash Point 26°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.398
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.8
Linear Formula C2H5CH(OH)CH3
Viscosity 4.21 mPa.s (20°C)
Beilstein 01, 373
Merck Index 15, 1543
Solubility Information Solubility in water: in water: 125g/L (20°C)
Formula Weight 74.12
Percent Purity 99%
Chemical Name or Material (S)-(+)-sec-Butanol, 0.99
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (S)-(+)-sec-Butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'