missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%

Brand:  Alfa Aesar™ L00577.14

Code : NEW

Additional Details : CAS Number : 22236-45-9 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11300124

  • £75.70 / 25g


Estimated Shipment date
from Supplier 19-01-2018
Add to basket

Description

Description

Applications
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in EtOH, THF, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
Specifications

Chemical Identifiers

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine
C9H13NO3S2
MFCD00013922
N-(p-Toluenesulfonyl)dimethylsulfoximine
22236-45-9
247.34
2119329

Specifications

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine
98%
247.34
2119329
167° to 169°C
22236-45-9
C9H13NO3S2
MFCD00013922
N-(p-Toluenesulfonyl)dimethylsulfoximine
25g
Videos
SDS