missing translation for 'onlineSavingsMsg'
Learn More

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, 98%, Thermo Scientific Chemicals

Catalog Number p-7058255
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
22236-45-9
C9H13NO3S2
247.327
MFCD00013922
IRNAWARRPQUZDU-UHFFFAOYSA-N
s,s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-p-tolylsulfonyl sulfoximine, dimethyl 4-methylbenzenesulfonyl imino-??-sulfanone, maybridge1_004531, s,s-dimethyl-n-p-tolylsulphonyl sulphoximide, dimethyl n-p-toluenesulfonyl sulfoximine, dimethyl-n-4-toluenesulfonyl sulfoximine, n-para-toluenesulfonyl dimethylsulfoximine, s.s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-4-methylphenylsulfonyl sulfoximide
99226
N-[dimethyl(oxo)-$l^{6}-sulfanylidene]-4-methylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)N=S(=O)(C)C
This item is not returnable. View return policy
22236-45-9
C9H13NO3S2
247.327
MFCD00013922
IRNAWARRPQUZDU-UHFFFAOYSA-N
s,s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-p-tolylsulfonyl sulfoximine, dimethyl 4-methylbenzenesulfonyl imino-??-sulfanone, maybridge1_004531, s,s-dimethyl-n-p-tolylsulphonyl sulphoximide, dimethyl n-p-toluenesulfonyl sulfoximine, dimethyl-n-4-toluenesulfonyl sulfoximine, n-para-toluenesulfonyl dimethylsulfoximine, s.s-dimethyl-n-p-toluenesulfonyl sulfoximine, s,s-dimethyl-n-4-methylphenylsulfonyl sulfoximide
99226
N-[dimethyl(oxo)-$l^{6}-sulfanylidene]-4-methylbenzenesulfonamide
CC1=CC=C(C=C1)S(=O)(=O)N=S(=O)(C)C

S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine, is used as methylene-transfer agent. The anion adds to carbonyl compounds to give epoxides which can react with a further mole of reagent with ring expansion to give 2,2-disubstituted oxetanes. It is also used as a pharmaceutical intermediate.

Solubility
Soluble in EtOH, THF, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 167°C to 169°C
Quantity 5 g
Beilstein 2119329
Solubility Information Soluble in EtOH,THF,DMSO.
Formula Weight 247.34
Percent Purity 98%
Chemical Name or Material S,S-Dimethyl-N-(p-toluenesulfonyl)sulfoximine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'