missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%, Thermo Scientific Chemicals

Catalog Number 10593884
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
93621-94-4
C8H17NO2
159.23
MFCD00210016
HDXYTIYVVNJFLU-YUMQZZPRSA-N
2s,5s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, 2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole
7019262
(2S,5S)-2,5-bis(methoxymethyl)pyrrolidine
COCC1CCC(N1)COC
This item is not returnable. View return policy
93621-94-4
C8H17NO2
159.23
MFCD00210016
HDXYTIYVVNJFLU-YUMQZZPRSA-N
2s,5s-2,5-bis methoxymethyl pyrrolidine, s,s-2,5-bis-methoxymethylpyrrolidine, s,s-2,5-bis methoxymethyl pyrrolidine, s,s-+ 2,5-bis methoxymethyl pyrrolidine, s,s-+-2,5-bis methoxymethyl pyrrolidine, 2s-2alpha,5beta-bis methoxymethyl pyrrolidine, 2s,5s---2,5-bis methoxymethyl pyrrolidine, 2s,5s-2,5-bis methoxymethyl-2,3,4,5-tetrahydropyrrole
7019262
(2S,5S)-2,5-bis(methoxymethyl)pyrrolidine
COCC1CCC(N1)COC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490
Quantity 1 g
Specific Rotation Condition +8° (20°C c=3,EtOH)
Specific Rotation + 8.00
Formula Weight 159.23
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material (S,S)-(+)-2,5-Bis(methoxymethyl)pyrrolidine, 96%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'