missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-[2-(4'-Isopropyloxazolin-2'yl)ferrocenyl]diphenylphosphine, 97%, Thermo Scientific Chemicals

Catalog Number p-4258879
Click to view available options
Quantity:
100 mg
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
163169-29-7
C28H28FeNOP
481.36
MFCD15144869
FELCHLOAEMNOQM-KISGOYOSNA-N
(S)[(Sp)-2-(Diphenylphosphino)ferrocenyl]-4-isopropyloxazoline
(S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine
[Fe].c1cccc1.CC(C)[C@H]1COC(=N1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
163169-29-7
C28H28FeNOP
481.36
MFCD15144869
FELCHLOAEMNOQM-KISGOYOSNA-N
(S)[(Sp)-2-(Diphenylphosphino)ferrocenyl]-4-isopropyloxazoline
(S,S)-[2-(4-Isopropyl-2-oxazolinyl)ferrocenyl]diphenylphosphine
[Fe].c1cccc1.CC(C)[C@H]1COC(=N1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 162.0°C to 165.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 481.35
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (S,S)-[2-(4′-Isopropyloxazolin-2′yl)ferrocenyl]diphenylphosphine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'