missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%

Catalog Number p-7028602
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1052707-27-3
C22H22Cl2N2
385.332
MFCD09265324
SHNGCXWOHADIKG-IXOXMDGESA-N
1s, 2s-1,2-di-1-naphthyl-ethylenediamine dihydrochloride, 1s,2s-1,2-di-1-naphthylethylenediamine dihydrochloride, 1s,2s-1,2-di-1-naphthalenyl-1,2-ethanediaminehydrochloride, 1s,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine dihydrochloride, s, s-1,2-bis 1-naphthyl-1,2-ethanediamine dihydrochloride, 1s,2s-1,2-bis naphthalen-1-yl ethane-1,2-diamine dihydrochloride, 1s,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine-hydrogen chloride 1/2, s,s-1,2-di 1-naphthyl-1,2-ethanediamine dihydrochloride, ee
57468034
(1S,2S)-1,2-dinaphthalen-1-ylethane-1,2-diamine;dihydrochloride
C1=CC=C2C(=C1)C=CC=C2C(C(C3=CC=CC4=CC=CC=C43)N)N.Cl.Cl
This item is not returnable. View return policy
1052707-27-3
C22H22Cl2N2
385.332
MFCD09265324
SHNGCXWOHADIKG-IXOXMDGESA-N
1s, 2s-1,2-di-1-naphthyl-ethylenediamine dihydrochloride, 1s,2s-1,2-di-1-naphthylethylenediamine dihydrochloride, 1s,2s-1,2-di-1-naphthalenyl-1,2-ethanediaminehydrochloride, 1s,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine dihydrochloride, s, s-1,2-bis 1-naphthyl-1,2-ethanediamine dihydrochloride, 1s,2s-1,2-bis naphthalen-1-yl ethane-1,2-diamine dihydrochloride, 1s,2s-1,2-di naphthalen-1-yl ethane-1,2-diamine-hydrogen chloride 1/2, s,s-1,2-di 1-naphthyl-1,2-ethanediamine dihydrochloride, ee
57468034
(1S,2S)-1,2-dinaphthalen-1-ylethane-1,2-diamine;dihydrochloride
C1=CC=C2C(=C1)C=CC=C2C(C(C3=CC=CC4=CC=CC=C43)N)N.Cl.Cl
Melting Point >200°C
Quantity 250 mg
Optical Rotation 235° (c=1 in Water)
Formula Weight 385.35
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (S,S)-1,2-Di(1-naphthyl)-1,2-ethanediamine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'