Learn More
Alfa Aesar™ (S,S)-1,2-Bis(2-methoxyphenyl)-1,2-ethanediamine, 97+%
Brand: Alfa Aesar™ H60623.03
Code : NEW
Additional Details : CAS Number : 148240-65-7 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
(S,S)-1,2-Bis(2-methoxyphenyl)-1,2-ethanediamine | |
C16H20N2O2 | |
MFCD17013980 |
148240-65-7 | |
272.34 |
Specifications
(S,S)-1,2-Bis(2-methoxyphenyl)-1,2-ethanediamine | |
≥97% | |
272.34 | |
1g |
148240-65-7 | |
C16H20N2O2 | |
MFCD17013980 |