missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S protein RBD anti-SARS-CoV-2, Clone: AM180, ACROBiosystems
HEK293 Monoclonal Antibody
Brand: ACROBiosystems SPD-M180-100ug
Description
This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested.Specifications
| S protein RBD | |
| AM180 | |
| Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization. | |
| 100 μg | |
| Primary | |
| SARS-CoV-2 | |
| Recombinant Antibodies | |
| IgG1 κ |
| ELISA | |
| Unconjugated | |
| Human (HEK293) | |
| RUO | |
| This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested. | |
| -20°C | |
| Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction