missing translation for 'onlineSavingsMsg'
Learn More

(S)-α-Propargylglycine, 98%, 98% ee, ACROS Organics™

Brand:  Acros Organics 441215000

Code : 29

Additional Details : CAS Number : 23235-01-0 Weight : 0.05050kg

Quantity 500mg

 View more versions of this product

Product Code. 11935371

  • £436.40 / 500mg


Estimated Shipment
25-06-2018
Add to basket

Specifications

Chemical Identifiers

(S)-α-Propargylglycine
23235-01-0
100.0
98%
113.12
98% ee
97.5
97.5 to 102.5%
C5H7NO2
(S)-2-Amino-4-pentynoic acid

Specifications

(S)-α-Propargylglycine
23235-01-0
100.0
Authentic
97.5 to 102.5%
C5H7NO2
(S)-2-Amino-4-pentynoic acid
White to pale yellow
98% ee
97.5
98.0% min.
Powder
98%
113.12
-31° to -35° (20°C, 589nm) (c=1 in water)
500mg
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • (S)-alpha-Propargylglycine

Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.

SDS