missing translation for 'onlineSavingsMsg'
Learn More

(S)-α-Propargylglycine, 98%, 98% ee, ACROS Organics™

Brand:  Acros Organics 441211000

Code : 29

Additional Details : CAS Number : 23235-01-0 Weight : 0.05010kg

Quantity 100mg
Packaging Glass bottle

 View more versions of this product

Product Code. 11925371

  • £124.40 / 100mg


EU Stock 1
Add to basket

Specifications

Chemical Identifiers

(S)-α-Propargylglycine
23235-01-0
100.0
98%
113.12
98% ee
97.5
97.5 to 102.5%
C5H7NO2
(S)-2-Amino-4-pentynoic acid

Specifications

(S)-α-Propargylglycine
23235-01-0
100.0
Authentic
97.5 to 102.5%
C5H7NO2
113.12
-31° to -35° (20°C, 589nm) (c=1 in water)
100mg
98% ee
97.5
98.0% min.
Powder
98%
Glass bottle
(S)-2-Amino-4-pentynoic acid
White to pale yellow
Videos
SDS