missing translation for 'onlineSavingsMsg'
Learn More

(S)-α-Propargylglycine, 98%, 98% ee, Thermo Scientific™

Catalog Number p-4259056
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
23235-01-0
C5H7NO2
113.12
MFCD00077855
DGYHPLMPMRKMPD-BYPYZUCNSA-N
l-propargylglycine, s-2-aminopent-4-ynoic acid, 2s-2-aminopent-4-ynoic acid, 2s-2-amino-4-pentynoic acid, s-2-amino-4-pentynoic acid, unii-8r9u8rvt6v, 8r9u8rvt6v, 4-pentynoic acid, 2-amino-, 2s, l-2-propynylglycine, s-propargylglycine
168091
(2S)-2-aminopent-4-ynoic acid
N[C@@H](CC#C)C(O)=O
This item is not returnable. View return policy
23235-01-0
C5H7NO2
113.12
MFCD00077855
DGYHPLMPMRKMPD-BYPYZUCNSA-N
l-propargylglycine, s-2-aminopent-4-ynoic acid, 2s-2-aminopent-4-ynoic acid, 2s-2-amino-4-pentynoic acid, s-2-amino-4-pentynoic acid, unii-8r9u8rvt6v, 8r9u8rvt6v, 4-pentynoic acid, 2-amino-, 2s, l-2-propynylglycine, s-propargylglycine
168091
(2S)-2-aminopent-4-ynoic acid
N[C@@H](CC#C)C(O)=O
CAS Min % 97.5
CAS Max % 100.0
Melting Point >180°C
Color White to Yellow
Enantiomeric Excess 98.0% min.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation -32
Formula Weight 113.12
Percent Purity 98%,98% ee
Physical Form Powder
Chemical Name or Material (S)-α-Propargylglycine, 98% ee
Show More Show Less
Product Identifier
  • (S)-alpha-Propargylglycine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'