missing translation for 'onlineSavingsMsg'
Learn More

(S)-Phenyl superquat, 98%, ACROS Organics™

Brand:  Acros Organics 337810010

Code : 29

Additional Details : CAS Number : 168297-84-5 Weight : 0.05100kg

Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10285631

  • £71.00 / 1g


Estimated Shipment
24-05-2018
Add to basket

Specifications

Chemical Identifiers

(S)-Phenyl superquat
168297-84-5
100.0
C11H13NO2
MFCD00274262
98%
97.5
98%
191.23
(S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone

Specifications

(S)-Phenyl superquat
98%
97.5
100.0
C11H13NO2
MFCD00274262
1g
168297-84-5
Glass bottle
98%
191.23
(S)-5, 5-Dimethyl-4-phenyl-2-oxazolidinone
Videos
SDS