missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Niguldipine hydrochloride, Tocris Bioscience™

α1 antagonist, L-type Ca2+ channel blocker

Brand:  Tocris (Bio-Techne) 112310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13448603

  • £139.65 / 10mg

Estimated Shipment date
from Supplier 21-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

L-type Ca2+ channel blocker and α1A-adrenoceptor antagonist; more active enantiomer.

R-enantiomer also available.

Specifications

Chemical Identifiers

113145-69-0
646.181
s +-niguldipine hydrochloride, unii-1g8c7qs9sr, s-niguldipine hydrochloride, s-+-niguldipine hydrochloride, 1g8c7qs9sr, niguldipine hydrochloride, +-niguldipine hydrochloride, r---niguldipine hydrochloride, s +-1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 3-4,4-diphenyl-1-piperidinyl-propyl methyl ester hydrochloride
5-O-[3-(4,4-diphenylpiperidin-1-yl)propyl] 3-O-methyl (4S)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
C36H40ClN3O6
MHOSUIMBPQVOEU-WAQYZQTGSA-N
16219720
CC1=C(C(C(=C(N1)C)C(=O)OCCCN2CCC(CC2)(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl

Specifications

10mg
113145-69-0
s +-niguldipine hydrochloride, unii-1g8c7qs9sr, s-niguldipine hydrochloride, s-+-niguldipine hydrochloride, 1g8c7qs9sr, niguldipine hydrochloride, +-niguldipine hydrochloride, r---niguldipine hydrochloride, s +-1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 3-4,4-diphenyl-1-piperidinyl-propyl methyl ester hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OCCCN2CCC(CC2)(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl
646.181
646.18
(S)-(+)-Niguldipine hydrochloride
C36H40ClN3O6
MHOSUIMBPQVOEU-WAQYZQTGSA-N
5-O-[3-(4,4-diphenylpiperidin-1-yl)propyl] 3-O-methyl (4S)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
16219720
>98%
Videos
Documents