missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-Niguldipine hydrochloride, Tocris Bioscience™

α1 antagonist, L-type Ca2+ channel blocker

Brand:  Tocris (Bio-Techne) 112350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13458603

  • £414.72 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

L-type Ca2+ channel blocker and α1A-adrenoceptor antagonist; more active enantiomer. (R)-(-)-Niguldipine hydrochloride (Cat. No. 1124) also available.
Specifications

Chemical Identifiers

113145-69-0
646.181
s +-niguldipine hydrochloride, unii-1g8c7qs9sr, s-niguldipine hydrochloride, s-+-niguldipine hydrochloride, 1g8c7qs9sr, niguldipine hydrochloride, s-+-niguldipine hydrochloride, +-niguldipine hydrochloride, r---niguldipine hydrochloride, s +-1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 3-4,4-diphenyl-1-piperidinyl-propyl methyl ester hydrochloride
5-O-[3-(4,4-diphenylpiperidin-1-yl)propyl] 3-O-methyl (4S)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
C36H40ClN3O6
MHOSUIMBPQVOEU-WAQYZQTGSA-N
16219720
CC1=C(C(C(=C(N1)C)C(=O)OCCCN2CCC(CC2)(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl

Specifications

(S)-(+)-Niguldipine hydrochloride
113145-69-0
s +-niguldipine hydrochloride, unii-1g8c7qs9sr, s-niguldipine hydrochloride, s-+-niguldipine hydrochloride, 1g8c7qs9sr, niguldipine hydrochloride, s-+-niguldipine hydrochloride, +-niguldipine hydrochloride, r---niguldipine hydrochloride, s +-1,4-dihydro-2,6-dimethyl-4-3-nitrophenyl-3,5-pyridinedicarboxylic acid 3-4,4-diphenyl-1-piperidinyl-propyl methyl ester hydrochloride
CC1=C(C(C(=C(N1)C)C(=O)OCCCN2CCC(CC2)(C3=CC=CC=C3)C4=CC=CC=C4)C5=CC(=CC=C5)[N+](=O)[O-])C(=O)OC.Cl
646.181
646.18
50mg
C36H40ClN3O6
MHOSUIMBPQVOEU-WAQYZQTGSA-N
5-O-[3-(4,4-diphenylpiperidin-1-yl)propyl] 3-O-methyl (4S)-2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
16219720
>98%
Videos